EMAIL THIS PAGE TO A FRIEND
Journal of investigative medicine : the official publication of the American Federation for Clinical Research
Isoflurane provides neuroprotection in neonatal hypoxic ischemic brain injury.
PMID 23884213
Abstract
Isoflurane is a volatile anesthetic that is widely used clinically as an inhalational anesthetic. In recent years, several studies have indicated that isoflurane has neuroprotective properties. This has led to the beneficial effects of isoflurane being analyzed in both cell culture and animal models, including various models of brain injury. Neonatal hypoxia ischemia may be characterized as injury that occurs in the immature brain, resulting in delayed cell death via excitotoxicity and oxidative stress. These adverse events in the developing brain often lead to detrimental neurological defects in the future. Currently, there are no well-established effective therapies for neonatal hypoxia ischemia. In line with this, isoflurane, which displays neuroprotective properties in several paradigms and has been shown to improve neurological deficits caused by brain injuries, has the capability to be an extremely relevant clinical therapy for the resolution of deficits concomitant with neonatal hypoxic ischemic brain injuries. This review therefore seeks to explore and analyze the current information on isoflurane, looking at general isoflurane anesthetic properties, and the protection it confers in different animal models, focusing particularly on neuroprotection as shown in studies with neonatal hypoxic ischemic brain injury.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
792632
Isoflurane
C3H2ClF5O