EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Copper-catalyzed nucleophilic trifluoromethylation of propargylic halides.
PMID 23884450
Abstract
Reactions of propargylic halides with trifluoromethyltrimethylsilane in the presence of a catalytic amount of copper(I) thiophene-2-carboxylate (CuTC) have been found to give the corresponding trifluoromethylated products in good to high yields with a high selectivity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T32603
2-Thiophenecarboxylic acid, ReagentPlus®, 99%
C5H4O2S