EMAIL THIS PAGE TO A FRIEND
Asian Pacific journal of cancer prevention : APJCP
Serum talin-1 is a potential novel biomarker for diagnosis of hepatocellular carcinoma in Egyptian patients.
PMID 23886189
Abstract
Hepatocellular carcinoma (HCC) is a major cause of cancer mortality worldwide. The outcome of HCC depends mainly on its early diagnosis. To date, the performance of traditional biomarkers is unsatisfactory. Talins were firstly identified as cytoplasmic protein partners of integrins but Talin-1 appears to play a crucial role in cancer formation and progression. Our study was conducted to assess the diagnostic value of serum Talin-1 (TLN1) compared to the most feasible traditional biomarker alpha-fetoprotein (AFP) for the diagnosis of HCC. TLN1 was detected using enzyme linked immunosorbent assay (ELISA) in serum samples from 120 Egyptian subjects including 40 with HCC, 40 with liver cirrhosis (LC) and 40 healthy controls (HC). ROC curve analysis was used to create a predictive model for TLN1 relative to AFP in HCC diagnosis. Serum levels of TLN1 in hepatocellular carcinoma patients were significantly higher compared to the other groups (p<0.0001). The diagnostic accuracy of TLN1 was higher than that of AFP regarding sensitivity, specificity, positive predictive value and negative predictive value in diagnosis of HCC. The present study showed for the first time that Talin-1 (TLN1) is a potential diagnostic marker for HCC, with a higher sensitivity and specificity compared to the traditional biomarker AFP.