EMAIL THIS PAGE TO A FRIEND
Journal of pediatric hematology/oncology
Biochemical, radiologic, ultrastructural, and genetic evaluation of iron overload in acute leukemia and iron-chelation therapy.
PMID 23887025
Abstract
Iron overload in hereditary hemochromatosis and hematologic malignancy has unfavorable effects on morbidity. Herein, 53 children (age 108.4±58.3 mo, 25 girls and 28 boys) with acute myeloblastic and lymphoblastic leukemia, who received 4 different chemotherapy protocols, were evaluated for iron overload throughout chemotherapy. Iron overload arose: (1) before chemotherapy, which was dependent on neither chemotherapy nor packed red blood cell transfusions and (2) after chemotherapy, which was dependent on the duration and nature of chemotherapy and partially on transfusion of packed red blood cells. Iron overload was documented in 75% of patients with a ferritin level >1000 ng/mL, by liver and heart magnetic resonance imaging, and they were administered iron-chelation therapy with success. Three of 10 radiologically iron-overloaded patients were heterozygous for H63D mutation. Aminolevulinic acid and porphobilinogen levels were normal. Light microscopic examination of the bone marrow revealed increased iron granules in erythroblasts, platelets, and megakaryocytes, iron-laden macrophages, free iron in the matrix, dyshematopoiesis, and apoptotic cells. Electron microscopic examination revealed iron-laden secondary lysosomes and autolysosomes in normoblasts and iron-laden primary granules in promyelocytes, irrelevant to the ferritin level, implying autophagia due to chemotherapy as a source of the excess iron. We think that iron overload, which is an important complication of acute leukemia, should be evaluated separately from "transfusion overload," and the management principles specific to leukemia should be implemented.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P1134
Porphobilinogen, powder
C10H14N2O4