EMAIL THIS PAGE TO A FRIEND
Current pain and headache reports
Update on neuropathic pain treatment: ion channel blockers and gabapentinoids.
PMID 23888370
Abstract
Neuropathic pain is a debilitating chronic pain condition, which remains difficult to treat. The current mainstays of treatment include physical therapy, interventional procedures and medications. Among medications, ion channel blockers and gabapentinoids are the 2 classes of drugs commonly used to treat neuropathic pain. It has been suggested that these medications may be useful to treat a variety of neuropathic pain conditions. This article provides several updates on the utility of both ion channel blockers and gabapentinoids for the treatment of neuropathic pain.