EMAIL THIS PAGE TO A FRIEND
Revue medicale de Liege
[Is the concept of psychological dependence still valid today?].
PMID 23888575
Abstract
Historically, psychological dependence to alcohol has been dissociated from physical dependence. Progress in neurosciences and genetics now question this dichotomy as complex neurobiological mechanisms and genetic factors seem to underlie craving, which is typically considered as the clinical manifestation of psychological dependence.