EMAIL THIS PAGE TO A FRIEND
Revue medicale de Liege
[Rapid opiate detoxification under anesthesia (RODA)].
PMID 23888580
Abstract
Rapid Opiate Detoxification under Anesthesia (RODA) involves the use of opiate antagonists combined with anesthesia and pharmacotherapy to reduce withdrawal symptoms. The aim of our study was to measure the plasma concentrations of heroin metabolites and methadone during anesthesia and patient stay at the hospital in order to assess the amount of active substances at each protocol step. Plasma concentrations of antagonists were also quantified and compared to the recommended target values. Blood samples were drawn in 10 patients undergoing RODA at different times of the procedure (during anesthesia, in post-anesthesia care unit and in psychiatry unit). The plasma concentrations of heroin metabolites, methadone and antagonists were measured using a previously described method. Heroin active metabolites were no longer detected in the patient blood when helshe left the hospital; by contrast, methadone was still present at significant concentrations 3 days after the beginning of the detoxification procedure. Naltrexone analysis allowed us to adjust doses to insure opiate receptor blockade during acute withdrawal, which is a critical period.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M0267
(±)-Methadone hydrochloride, powder, ≥98%
C21H27NO · HCl
M-019
(±)-Methadone solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material
C21H27NO
M-007
(±)-Methadone solution, 1 mg/mL in methanol, ampule of 1 mL, certified reference material
C21H27NO
M3528
Morphine 6-β-D-glucuronide hydrate, ≥90% (HPLC)
C23H27NO9 · 2H2O
M-005
Morphine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material
C17H19NO3
M-030
Morphine solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material
C17H19NO3
M8777
Morphine sulfate salt pentahydrate
C34H40N2O10S · 5H2O
M9524
Morphine sulfate salt solution, 1.0 mg/mL in methanol, drug standard
C34H38N2O6 · H2SO4
M-018
Morphine-3-β-D-glucuronide solution, 100 μg/mL in methanol: water (1:1), ampule of 1 mL, certified reference material
C23H27NO9
M-031
Morphine-3-β-D-glucuronide solution, 1 mg/mL in methanol with 0.05% NaOH, ampule of 1 mL, certified reference material
C23H27NO9
M-046
Morphine-6-β-D-glucuronide solution, 1.0 mg/mL in methanol: water (2:8), ampule of 1 mL, certified reference material
C23H27NO9
M-096
Morphine-6-β-D-glucuronide solution, 100 μg/mL in methanol: water (1:1), ampule of 1 mL, certified reference material
C23H27NO9
N7758
Naloxone hydrochloride dihydrate, ≥98% (TLC and titration), powder
C19H21NO4 · HCl · 2H2O
N-004
Naloxone solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material
C19H21NO4
N-007
Naltrexone solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material
C20H23NO4