EMAIL THIS PAGE TO A FRIEND
Klinichna khirurhiia
[Hepatic dysfunction correction in patients with obturation jaundice].
PMID 23888709
Abstract
Comparative analysis of results of examination and treatment of 54 patients, suffering obturation jaundice syndrome, is presented. The presence and severity of hepatic dysfunction was determined in accordance to indices of cytolysis and cholestasis syndromes, and its severity--in accordance to V. P. Zinevich criteria (1986). There was established, that timely elimination of obturation jaundice, using miniinvasive interventions and subsequent conduction of complex hepatotropic therapy, using L-ornitin L-aspartate, permits to correct hepatic dysfunction effectively.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
O7125
L-Ornithine L-aspartate salt, powder
C9H19N3O6