EMAIL THIS PAGE TO A FRIEND
Environmental toxicology and pharmacology
Interactive effect of oxytetracycline and lead on soil enzymatic activity and microbial biomass.
PMID 23892283
Abstract
Interactive effect of oxytetracyline (OTC) and lead on soil enzymatic activity and population of microbes was studied in the paper. The results showed effect of pollutants on bacteria, actinomycetes and enzymatic activity increased in the order: (OTC+Pb)>Pb>OTC, (OTC+Pb)>Pb>OTC and (OTC+Pb)>OTC>Pb, respectively. However, impact of pollutants on fungi decreased in the order: (OTC+Pb)