EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Heating under high-frequency inductive conditions: application to the continuous synthesis of the neurolepticum olanzapine (Zyprexa).
PMID 23893845