EMAIL THIS PAGE TO A FRIEND
Blood
Retinoic acid plus arsenic trioxide, the ultimate panacea for acute promyelocytic leukemia?
PMID 23894153
Abstract
Rarely in the field of cancer treatment did we experience as many surprises as with acute promyelocytic leukemia (APL). Yet, the latest clinical trial reported by Lo-Coco et al in the New England Journal of Medicine is a practice-changing study, as it reports a very favorable outcome of virtually all enrolled low-intermediate risk patients with APL without any DNA-damaging chemotherapy. Although predicted from previous small pilot studies, these elegant and stringently controlled results open a new era in leukemia therapy.