EMAIL THIS PAGE TO A FRIEND
Tissue Engineering: Part A
Bone marrow-derived mesenchymal stem cells become antiangiogenic when chondrogenically or osteogenically differentiated: implications for bone and cartilage tissue engineering.
PMID 23895198