EMAIL THIS PAGE TO A FRIEND
Annals of the Rheumatic Diseases
Alcohol consumption as a preventive factor for developing rheumatoid arthritis: a dose-response meta-analysis of prospective studies.
PMID 23897767