EMAIL THIS PAGE TO A FRIEND
Drug testing and analysis
Discrimination of cathinone regioisomers, sold as 'legal highs', by Raman spectroscopy.
PMID 23900999
Abstract
The discrimination of a cross section of cathinone regioisomers, sold as 'legal highs', using Raman spectroscopy, is reported here. Mephedrone and flephedrone were identified in 'legal high' products sold in Irish head shops, and their 2, 3 and 4-isomers were synthesized as reference standards. The 3,4-methylenedioxy substituted cathinones, methylone, butylone and methylenedioxypyrovalerone (MDPV), were also identified in 'legal highs' and their 2,3-isomers were synthesized for comparison. In addition, alpha- and beta-naphyrone were synthesized. Raman spectra of all the isomers were obtained using far-red excitation (785 nm) and it was found possible to discriminate the isomers of each substituted cathinone. In addition, Raman spectra were also recorded for a number of head shop products and, by comparison with the reference standards, correct isomer assignment for 4-mephedrone, 3-flephedrone, 3,4-methylone, 3,4-butylone, 3,4-MDPV, alpha-naphyrone and beta-naphyrone was achieved, thus providing a non-destructive, high-throughput and minimal sample preparation technique for the discrimination of such drug isomers.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
93784
Caffeine, PharmaGrade, EP, Manufactured under appropriate GMP controls for pharma or biopharmaceutical production
C8H10N4O2
C0750
Caffeine, powder, ReagentPlus®
C8H10N4O2
C8960
Caffeine, BioXtra
C8H10N4O2
C1778
Caffeine, Sigma Reference Standard, vial of 250 mg
C8H10N4O2
C0100000
Caffeine, European Pharmacopoeia (EP) Reference Standard
C8H10N4O2
C7731
Caffeine, meets USP testing specifications, anhydrous
C8H10N4O2
1085003
Caffeine, United States Pharmacopeia (USP) Reference Standard
C8H10N4O2
PHR1009
Caffeine, Pharmaceutical Secondary Standard; Certified Reference Material
C8H10N4O2
56396
Caffeine, certified reference material, TraceCERT®
C8H10N4O2
05-0370
Caffeine, SAJ special grade, ≥98.5%
C8H10N4O2
Y0000787
Caffeine for system suitability, European Pharmacopoeia (EP) Reference Standard
C8H10N4O2
1086006
Caffeine melting point standard, United States Pharmacopeia (USP) Reference Standard
C8H10N4O2
PHR1095
Caffeine Melting Point Standard, Pharmaceutical Secondary Standard; Certified Reference Material
C8H10N4O2
L7757
Lidocaine, powder
C14H22N2O
L1026
Lidocaine, analytical standard
C14H22N2O
PHR1034
Lidocaine, Pharmaceutical Secondary Standard; Certified Reference Material
C14H22N2O
L0595000
Lidocaine, European Pharmacopoeia (EP) Reference Standard
C14H22N2O
1366002
Lidocaine, United States Pharmacopeia (USP) Reference Standard
C14H22N2O
41019
Melting point standard 235-237°C, analytical standard
C8H10N4O2
75035
Mettler-Toledo Calibration substance ME 18872, Caffeine, analytical standard, for the calibration of the thermosystem 900, traceable to primary standards (LGC)
C8H10N4O2