EMAIL THIS PAGE TO A FRIEND
Journal of leukocyte biology
Technical advance: Inhibition of neutrophil chemotaxis by colchicine is modulated through viscoelastic properties of subcellular compartments.
PMID 23901122
Abstract
Colchicine is an efficient drug for the management of inflammatory diseases, such as gouty arthritis and familial Mediterranean fever. It affects neutrophil activity by interfering with the formation of microtubules. To test the hypothesis that therapeutic concentrations of colchicine modulate the mechanical properties of these cells, we applied a combination of biophysical techniques (optical stretching and microrheology) to analyze cellular deformability. The contribution of the subcellular compartments to the regulation of cell mechanics was determined by fitting a multicomponent model of cellular viscoelasticity to time-dependent deformation curves. Neutrophils were found to be less deformable in response to 10 ng/ml colchicine. The model-based analysis of cellular deformation revealed a decrease in cytoplasmatic elasticity and a substantial increase in both elasticity and viscosity of the cell membrane compartment in response to colchicine. These results correlate with a reduced number of cytoplasmatic microtubules and an increase in subcortical actin filaments. The latter finding was confirmed by microrheology and fluorescence microscopy. Neutrophil migration through small pores requiring substantial cellular deformations, but not through large pores, was significantly impaired by colchicine. These data demonstrate that colchicine determines mechanics of neutrophils and, thereby, motility in confined spaces, which is crucial during extravasation of neutrophils in response to inflammatory stimuli.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Y0000209
Colchicine for system suitability, European Pharmacopoeia (EP) Reference Standard
C22H25NO6