EMAIL THIS PAGE TO A FRIEND
Journal of nanoscience and nanotechnology
Preparation and photocatalytical performance of TiO2:SiO2 nanocomposites produced by the polymeric precursors method.
PMID 23901540
Abstract
Anatase TiO2 is a promising photocatalyst due to its chemical stability, non-toxic characteristics, notable UV light absorption as well as photo-corrosion resistance and oxidative properties. Surface area and TiO2 dispersion quality are important factors that affect photoactivity of TiO2:SiO2 nanocomposites. In order to improve these factors, TiO2 nanoparticles were immobilized on mesoporous silica substrate through the polymeric precursors method, obtaining the nanocomposites in a simple routine. The TiO2 resin was synthesized by the polymeric precursors method and different resin thickness (0.5; 1.0; 2.0; 3.5; 5.0 nm) on silica were synthesized by calcination during 4 hours at 450 degrees C in pH 1.5. The selected pH for immobilization ensured adhesion of TiO2 nanoparticles onto the silica substrate surface. X-Ray Diffraction patterns indicate that all samples were predominantly anatase phase and immobilization improved surface area. Ametryn kinetic evaluation presents better results for SAM 3.5 and SAM 0.5. The results show that difference in TiO2 loading, surface area and crystallinity of samples are factors that influence photocatalytic efficiency.