EMAIL THIS PAGE TO A FRIEND
Journal of clinical and diagnostic research : JCDR
Simultaneous determination of levodopa and carbidopa from fava bean, green peas and green beans by high performance liquid gas chromatography.
PMID 23905090
Abstract
According to many studies, sprouted fava beans are a rich source of levo-dihydroxy phenylalanine (L-dopa) the precursor of dopamine, and they are now being investigated for use in the management of Parkinson's disease. The addition of Carbidopa (C-dopa) can reduce the daily use of the L-dopa dosage requirements and it can also reduce the side effects which are associated with the L-dopa administration. The present research was conducted to find the levo-dihydroxy phenylalanine (L-dopa) and Carbidopa (C-dopa) in fava beans, green peas and green beans by High Performance Gas Chromatography (HPLC). Carbidopa (C-dopa) is a peripheral decarboxylase inhibitor. As a substitution therapy, it used in combination to treat Parkinson's disease. We obtained L-dopa and C-dopa from fava beans which were in the fresh and dry sprouted form, whose concentrations were 1.4,1.5 and 2.6,2.4 mg/ml respectively. The maximal stimulation of the L-DOPA content was seen on day 8 for the fava beans, which was 100% higher than that of the control level. The results of this study indicate that faba beans are a good source of natural L-dopa and C-dopa. The quantification of this capacity according to the stage and the plant part could be suitable for applications in the food industry and in plant medicine. The consumption of fava beans can increase the levels of L-dopa and C-dopa in the blood, with a marked improvement in the motor performance of the patients with parkinson disease, without any side effects.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
1095506
Carbidopa, United States Pharmacopeia (USP) Reference Standard
C10H14N2O4 · H2O