EMAIL THIS PAGE TO A FRIEND
Hepatology Research : the Official Journal of the Japan Society of Hepatology
Interleukin-21 mediates hepatitis B virus-associated liver cirrhosis by activating hepatic stellate cells.
PMID 23905760