EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
Urea-catalyzed construction of oxazinanes.
PMID 23907178
Abstract
Highly functionalized oxazinanes are efficiently prepared through urea-catalyzed formal [3 + 3] cycloaddition reactions of nitrones and nitrocyclopropane carboxylates. The reaction system is general with respect to both the nitrocyclopropane carboxylates and nitrones enabling the preparation of a large family of oxazinanes, typically in high yield. This method affords access to enantioenriched oxazinane products through chirality transfer from enantioenriched nitrocyclopropane carboxylates.
Related Materials