EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Simultaneous enhanced diagnosis and photodynamic therapy of photosensitizer-doped perylene nanoparticles via doping, fluorescence resonance energy transfer, and antenna effect.
PMID 23907237
Abstract
We demonstrate a new strategy for preparing photosensitizer-doped perylene nanoparticles via doping, fluorescence resonance energy transfer, and antenna effect for simultaneous enhanced diagnosis and photodynamic therapy.