EMAIL THIS PAGE TO A FRIEND
Materials science & engineering. C, Materials for biological applications
Nanocomposites of rice and banana flours blend with montmorillonite: partial characterization.
PMID 23910294
Abstract
Rice and banana flours are inexpensive starchy materials that can form films with more improved properties than those made with their starch because flour and starch present different hydrophobicity. Montmorillonite (MMT) can be used to further improve the properties of starch-based films, which has not received much research attention for starchy flours. The aim of this work was to evaluate the mechanical and barrier properties of nanocomposite films of banana and rice flours as matrix material with addition of MMT as a nanofiller. MMT was modified using citric acid to produce intercalated structures, as verified by the X-ray diffraction pattern. The intercalated MMT was blended with flour slurries, and films were prepared by casting. Nanocomposite films of banana and rice flours presented an increase in the tensile at break and elongation percentage, respectively, more than their respective control films without MMT. This study showed that banana and rice flours could be alternative raw materials to use in making nanocomposite films.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
285234 Bentonite
682659 Nanoclay, hydrophilic bentonite