EMAIL THIS PAGE TO A FRIEND
The American journal of emergency medicine
A new and rapid method for epistaxis treatment using injectable form of tranexamic acid topically: a randomized controlled trial.
PMID 23911102
Abstract
Epistaxis is a common problem in the emergency department (ED). Sixty percent of people experience it at least once in their life. There are different kinds of treatment for epistaxis. This study intended to evaluate the topical use of injectable form of tranexamic acid vs anterior nasal packing with pledgets coated with tetracycline ointment. Topical application of injectable form of tranexamic acid (500 mg in 5 mL) was compared with anterior nasal packing in 216 patients with anterior epistaxis presented to an ED in a randomized clinical trial. The time needed to arrest initial bleeding, hours needed to stay in hospital, and any rebleeding during 24 hours and 1 week later were recorded, and finally, the patient satisfaction was rated by a 0-10 scale. Within 10 minutes of treatment, bleedings were arrested in 71% of the patients in the tranexamic acid group, compared with 31.2% in the anterior nasal packing group (odds ratio, 2.28; 95% confidence interval, 1.68-3.09; P < .001). In addition, 95.3% in the tranexamic acid group were discharged in 2 hours or less vs 6.4% in the anterior nasal packing group (P < .001). Rebleeding was reported in 4.7% and 11% of patients during first 24 hours in the tranexamic acid and the anterior nasal packing groups, respectively (P = .128). Satisfaction rate was higher in the tranexamic acid compared with the anterior nasal packing group (8.5 ± 1.7 vs 4.4 ± 1.8, P < .001). Topical application of injectable form of tranexamic acid was better than anterior nasal packing in the initial treatment of idiopathic anterior epistaxis.