EMAIL THIS PAGE TO A FRIEND
Experimental parasitology
In vivo evaluation of the efficacy of albendazole sulfoxide and albendazole sulfoxide loaded solid lipid nanoparticles against hydatid cyst.
PMID 23912040
Abstract
Cystic echinococcosis (CE) is caused by the larval stage of Echinococcus granulosus, which in this disease the metacestode develop in visceral organs especially liver and lungs. The disease is present worldwide and affects humans as well as herbivores including cattle, sheep, camels, horses and others. Benzimidazole carbamate derivatives, such as mebendazole and albendazole, are currently used for chemotherapeutic treatment of CE in inoperable patients and have to be applied in high doses for extended periods of time, and therefore adverse side effects are frequently observed. This study was designed to evaluate and compare the in vivo effects of 0.5 mg/kg, BID, albendazole sulfoxide (ricobendazole) and two different therapeutic regimens of 0.5 mg/kg BID and 2 mg/kg every 48 h of albendazole sulfoxide loaded solid lipid nanoparticles. Albendazole sulfoxide loaded solid lipid nanoparticles was prepared by solvent diffusion-evaporation method. Fifty Balb/c mice were infected by intraperitoneal injection of protoscoleces and 8 months post infection, the infected mice were treated for 15 days with the above mentioned regimens. They were then euthanized and the size and weight of the cysts as well as their ultrastructural changes were investigated. Although the cysts showed reduced size and weight in the treated animals but these reductions were not statistically significant. The cysts in the animals which received albendazole sulfoxide loaded SLN every 48 h showed more ultrastructural modification. However, these ultrastructural changes should be supported by further biochemical and molecular studies before introducing it as an efficient therapeutic regimen for treatment of human and animal hydatid disease.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
35395
Albendazole sulfoxide, VETRANAL®, analytical standard
C12H15N3O3S