EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Electrochemical and chemical tuning of the surface wettability of tetrathiafulvalene self-assembled monolayers.
PMID 23912526
Abstract
The tuning of the surface wettability of self-assembled monolayers of two novel tetrathiafulvalenes has been achieved by electrochemical and chemical means.