EMAIL THIS PAGE TO A FRIEND
International journal of pharmaceutics
A potential carrier based on liquid crystal nanoparticles for ophthalmic delivery of pilocarpine nitrate.
PMID 23916822
Abstract
Poor corneal penetration and short preocular retention of a clinical hydrophilic drug, pilocarpine nitrate (PN), for the treatment of open-angle glaucoma and acute angle-closure glaucoma, limit its ocular application. The purpose of this study was to investigate the potential of liquid crystal nanoparticles (LCNPs) for ocular delivery of PN. LCNPs were developed by a top-down method using glyceryl monoolein (GMO) and water in the presence of stabilizer Poloxamer 407. They were characterized by transmission electron microscopy (TEM) and small angle X-ray diffraction (SAXS). The size of LCNP is 202.28±19.32 nm and the encapsulation efficiency reached 61.03%. The in vitro release profiles indicated that PN could keep sustained release from PN-loaded LCNPs for 8h. An ex vivo corneal permeation study revealed that the apparent permeability coefficient of PN-loaded LCNPs was 2.05-fold higher than that of commercial eye drops. In addition, the topical administration test showed that PN-loaded LCNPs had a prolonged effect on decreasing intraocular pressure (IOP) of rabbits compared with commercial drug and physiological saline. In conclusion, LCNPs had been demonstrated to be potential for controlled-release ocular drug delivery.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
1538505 Pilocarpine, United States Pharmacopeia (USP) Reference Standard
C11H16N2O2