EMAIL THIS PAGE TO A FRIEND
Acta poloniae pharmaceutica
In vitro anti-oxidative activities and GC-MS analysis of various solvent extracts of Cassia singueana parts.
PMID 23923394
Abstract
The present study was conducted to investigate the anti-oxidative activities of different solvent extracts of Cassia singueana parts. Our results indicate that all the extracts have reducing power (Fe3+ --> Fe2+) and DPPH radical scavenging abilities. However, the ethyl acetate extract of the stem bark has the highest total reducing power whilst the ethanol extract of the stem bark has more potent free radical scavenging activity than all the other extracts. The ethyl acetate extract of the stem bark exhibited more powerful hydroxyl radical scavenging activity than other extracts whilst the aqueous extract of the leaves displayed more potent nitric oxide inhibition activity than other extracts. The GC-MS analysis of the ethyl acetate extract of the stem bark and the ethanol extract of the root and leaves indicated that several aromatic compounds, including phenolics, fatty acids, amino acids and triterpenoids were present in these extracts. Data from this study suggest that the parts of C. singueana possessed anti-oxidative activities and can be used as a potential alternative medicine for oxidative stress related non-communicable chronic diseases. Further experimental and clinical studies in this regard are warranted.