EMAIL THIS PAGE TO A FRIEND
Zhonghua er ke za zhi = Chinese journal of pediatrics
[Advances in the clinical and laboratory studies on methylmalonic aciduria combined with homocysteinemia type cblC].
PMID 23927810
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H1428000 Hydroxocobalamin, European Pharmacopoeia (EP) Reference Standard
C62H89CoN13O15P