EMAIL THIS PAGE TO A FRIEND
The Analyst
Suitable potentiometric enzyme sensors for urea and creatinine.
PMID 2393088
Abstract
Enzyme sensors for urea and creatinine were developed by coupling an ammonia gas-diffusion electrode with triacetate cellulose membranes entrapping urease or creatinine deiminase enzymes. Satisfactory results were obtained by using these sensors both in standard solutions and in authentic biological matrices.
Related Materials