EMAIL THIS PAGE TO A FRIEND
Journal of cosmetic science
Kinetics of inhibitory effect of isoferulic acid on mushroom tyrosinase.
PMID 23931087
Abstract
A study on the kinetics of inhibitory effect of isoferulic acid on the monophenolase and diphenolase activity of mushroom tyrosinase was carried out using enzymological kinetic analysis method in a Na2HPO4-NaH2PO4 buffer solution (pH = 6.8) at 30°C. It was found that isoferulic acid efficiently inhibits both monophenolase and diphenolase activities of mushroom tyrosinase under experimental conditions. Concentrations of isoferulic acid leading to 50% rate inhibition (IC50) on monophenolase and diphenolase activity were calculated to be 0.13 mmol/L and 0.39 mmol/L, respectively, which are much lower than that of arbutin (IC50 = 5.3 mmol/L for diphenolase activity). The presence of isoferulic acid also prolongs the lag period in the oxidation process of l-tyrosine via tyrosinase-a 4.3-min lagging was observed with the presence of 0.20 mmol/L isoferulic acid-compared to a 1.1-min lagging in the absence of isoferulic acid. The Lineweaver-Burk plot demonstrates a competitive behavior of isoferulic acid in the tyrosinase oxidation of l-3,4-dihydroxyphenylalanine, with maximum reaction rate (vm) and inhibition constant (KI) at 64.5 µM/min and 0.11 mmol/L, respectively.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
103012
3-Hydroxy-4-methoxycinnamic acid, predominantly trans, 97%
C10H10O4