EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Synthesis of 2-nitroglycals from glycals using the tetrabutylammonium nitrate-trifluoroacetic anhydride-triethylamine reagent system and base-catalyzed Ferrier rearrangement of acetylated 2-nitroglycals.
PMID 23931299
Abstract
A reagent system comprising tetrabutylammonium nitrate-trifluoroacetic anhydride-triethylamine has been developed for the synthesis of 2-nitroglycals from various protected glycals. The base-catalyzed Ferrier rearrangement on tri-O-acetylated 2-nitroglycals has been reported for the first time. Reactivity of these nitroacetates and associated selectivity has been examined, and some of the products have been converted into 2,3-diamino-2,3-dideoxyglycosides and methyl N-acetyl-D-lividosaminide.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
651664
Tetrabutylammonium azide
C16H36N4
86853
Tetrabutylammonium bisulfate, for ion pair chromatography, ≥99.0%
C16H37NO4S
86868
Tetrabutylammonium bisulfate, puriss., ≥99.0% (T)
C16H37NO4S
72453
Tetrabutylammonium bisulfate solution, ~55% in H2O
C16H37NO4S
98479
Tetrabutylammonium bisulfate solution, for ion pair chromatography, concentrate, ampule
C16H37NO4S
86857
Tetrabutylammonium bromide, for ion pair chromatography, ≥99.0%
C16H36BrN
426288
Tetrabutylammonium bromide, ACS reagent, ≥98.0%
C16H36BrN
193119
Tetrabutylammonium bromide, ReagentPlus®, ≥99.0%
C16H36BrN
462144
Tetrabutylammonium bromide solution, 50 wt. % in H2O
C16H36BrN
86852
Tetrabutylammonium chloride, for ion pair chromatography, ≥99.0%
C16H36ClN
86870
Tetrabutylammonium chloride, ≥97.0% (NT)
C16H36ClN
86862
Tetrabutylammonium chloride solution, for ion pair chromatography, concentrate, ampule
C16H36ClN
358665
Tetrabutylammonium cyanide, 95%
C17H36N2
79989
Tetrabutylammonium cyanide, technical, ≥80%
C17H36N2
216143
Tetrabutylammonium fluoride solution, 1.0 M in THF
C16H36FN
361399
Tetrabutylammonium fluoride solution, 75 wt. % in H2O
C16H36FN
155837
Tetrabutylammonium hydrogensulfate, 97%
C16H37NO4S
86854
Tetrabutylammonium hydroxide solution, ~40% in water, for ion chromatography
C16H37NO
86863
Tetrabutylammonium hydroxide solution, 54.0-56.0% in H2O
C16H37NO
86880
Tetrabutylammonium hydroxide solution, technical, ~40% in H2O (~1.5 M)
C16H37NO
178780
Tetrabutylammonium hydroxide solution, 40 wt. % in H2O
C16H37NO
230189
Tetrabutylammonium hydroxide solution, 1.0 M in methanol
C16H37NO
86903
Tetrabutylammonium iodide, for ion pair chromatography, ≥99.0%
C16H36IN
86912
Tetrabutylammonium iodide, for electrochemical analysis, ≥99.0%
C16H36IN
86890
Tetrabutylammonium iodide, ≥99.0% (AT)
C16H36IN
140775
Tetrabutylammonium iodide, reagent grade, 98%
C16H36IN
359173
Tetrabutylammonium nitrate, 97%
C16H36N2O3
86893
Tetrabutylammonium perchlorate, for electrochemical analysis, ≥99.0%
C16H36ClNO4
86885
Tetrabutylammonium perchlorate, ≥98.0% (T)
C16H36ClNO4
86833
Tetrabutylammonium phosphate monobasic, puriss., ≥99.0% (T)
C16H38NO4P