EMAIL THIS PAGE TO A FRIEND
International journal of antimicrobial agents
Detection of extended-spectrum β-lactamases and AmpC β-lactamases in Salmonella enterica isolates from patients in Denmark during 2008-2010.
PMID 23932741
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Y0000420 Cefotaxime acid, European Pharmacopoeia (EP) Reference Standard
C16H17N5O7S2