Description
Molecular Formula
Add to Cart
00268 Dextran, analytical standard, for GPC, 1,000
00269 Dextran, analytical standard, for GPC, 5,000
00270 Dextran, analytical standard, for GPC, 12,000
00271 Dextran, analytical standard, for GPC, 25,000
00891 Dextran, analytical standard, for GPC, 50,000
00892 Dextran, analytical standard, for GPC, 80,000
00893 Dextran, analytical standard, for GPC, 150,000
00894 Dextran, analytical standard, for GPC, 270,000
00895 Dextran, analytical standard, for GPC, 410,000
00896 Dextran, analytical standard, for GPC, 670,000
31430 Dextran, analytical standard, for GPC, Set Mp 1,000-400,000
31416 Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 1,000
31417 Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 5,000
31418 Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 12,000
31419 Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 25,000
31420 Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 50,000
31421 Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 80,000
31422 Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 150,000
31423 Dextran from Leuconostoc mesenteroides, for GPC, Mw 270,000
31424 Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 410,000
31425 Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 670,000
D8821 Dextran from Leuconostoc mesenteroides, average mol wt 60,000-76,000
09184 Dextran from Leuconostoc spp., Mr ~100,000
31387 Dextran from Leuconostoc spp., Mr 15,000-25,000
44886 Dextran from Leuconostoc spp., Mr ~70000
D8802 Dextran solution from Leuconostoc mesenteroides, 20 % (w/w) (Autoclaved)