EMAIL THIS PAGE TO A FRIEND
Zhongguo gu shang = China journal of orthopaedics and traumatology
[In vivo experimental study on MAO-ZK60 magnesium alloy bio-safety and degradation].
PMID 23937039
Abstract
To investigate the tissue toxicity and degradation of ZK60 magnesium alloy with micro-arc oxidation coatings (MAO-ZK60), in order to discuss the possibility of its potential application for orthopedic implantation. Eighteen Sprague-Dawley rats were randomized equally to three groups of A, B, C. MAO-ZK60 sticks were implanted in the femoral condyles of rats in group A (experimental group). Sticks of ZK60 magnesium alloy without any surface treatment (ZK60) were implanted in the femoral condyles of rats in group B (control group). The poly L-lactic acid (PLLA) sticks were implanted in the femoral condyles of rats in group C (control group). The changes of blood bio-chemical indexes of different groups were observed and compared. All the rats were sacrificed at 12 weeks and histological observation of liver and kidney were carried out to evaluate the hepatic and renal toxicity. Micro-CT was used to evaluate the degradation of the implants and to observe the bone-implant interface. GEHC MicroView software was operated to calculate the volume variation of magnesium alloy. There was no apparent biochemical index change with time in each group, and there was no significant change among each group. No significant pathology change of liver and kidney was detected among three groups. By using a micro-CT, a gap was found on the bone-implant interface at 4 weeks after implantation in group A, which decreased gradually at 8 weeks after implantation and continued to decrease at 12 weeks after implantation. A good combination between bone and implant formed at 12 weeks after implantation. Group A has less change of volume with time than group B (P < 0.05). ZK60 magnesium alloy with micro-arc oxidation coatings is safe in vivo. It has higher corrosion resistance than ZK60 magnesium alloy without any surface treatment.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
254118
Magnesium, chips, 6-35 mesh, 99.98% trace metals basis
Mg
474754
Magnesium, dendritic pieces, purified by distillation, 99.998% trace metals basis
Mg
465992
Magnesium, dendritic pieces, purified by distillation, 99.99% trace metals basis
Mg
254126
Magnesium, 20-230 mesh, reagent grade, 98%
Mg
63040
Magnesium, grit, ≥99.0% (KT)
Mg
253987
Magnesium, ReagentPlus®, powder, −50 mesh, ≥99%
Mg
465666
Magnesium, ReagentPlus®, powder, −325 mesh, 99.5% trace metals basis
Mg
13112
Magnesium, powder, ≥99%
Mg
266302
Magnesium, ReagentPlus®, ribbon, ≥99% trace metals basis
Mg
13103
Magnesium, ribbon, ≥99.5% Mg basis
Mg
299405
Magnesium, rod, diam. 6 mm, ≥99.9% trace metals basis
Mg
403148
Magnesium, turnings, ~1/8 in., 99.95% trace metals basis
Mg
200905
Magnesium, turnings, reagent grade, 98%
Mg
63035
Magnesium, purum, for Grignard reactions, ≥99.5%, turnings
Mg
723312
Magnesium, in a Sure/Seal bottle, turnings, 37.5 mmol
Mg
723320
Magnesium, in a Sure/Seal bottle, turnings, anhydrous tetrahydrofuran 37.5 mmol
Mg
735779
Magnesium, ingot, ≥99% trace metals basis
Mg
734365
Magnesium, wire, 127 μm diameter, 99.9% trace metals basis
Mg
735760
Magnesium, plate, 99.95%
Mg
499579
Magnesium preparation, Highly Reactive Rieke®
Metal, suspension, 2.5 g in THF
Mg