EMAIL THIS PAGE TO A FRIEND
The FEBS journal
Implications of enzyme deficiencies on mitochondrial energy metabolism and reactive oxygen species formation of neurons involved in rotenone-induced Parkinson's disease: a model-based analysis.
PMID 23937586
Abstract
Steadily growing experimental evidence suggests that mitochondrial dysfunction plays a key role in the age-dependent impairment of nerve cells underlying several neurodegenerative diseases. In particular, the citric acid cycle enzyme complex α-ketoglutarate dehydrogenase (KGDHC) and respiratory chain complex I of the respiratory chain often show reduced activities in the dopaminergic neurons involved in Parkinson's disease, both giving rise to an impaired mitochondrial energy metabolism as demonstrated in a number of in vitro studies with cell lines as well as isolated mitochondria. To understand the metabolic regulation underlying these experimental findings we used a detailed kinetic model of mitochondrial energy metabolism. First, we investigated the effect of complex I inhibition on energy production and formation of reactive oxygen species (ROS). Next, we applied the model to a situation where both KGDHC and complex I exhibit reduced activities. These calculations reveal synergistic effects with respect to the energy metabolism but antagonistic effects with respect to ROS formation: the drop in the ATP production capacity is more pronounced than at inhibition of either enzyme complex alone. Interestingly, however, the reduction state of the ROS-generating sites of the impaired complex I becomes significantly lowered if additionally the activity of the KGDHC is reduced. We discuss the pathophysiological consequences of these intriguing findings.