EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
The allosteric role of the AAA+ domain of ChlD protein from the magnesium chelatase of synechocystis species PCC 6803.
PMID 23940041
Abstract
Magnesium chelatase is an AAA(+) ATPase that catalyzes the first step in chlorophyll biosynthesis, the energetically unfavorable insertion of a magnesium ion into a porphyrin ring. This enzyme contains two AAA(+) domains, one active in the ChlI protein and one inactive in the ChlD protein. Using a series of mutants in the AAA(+) domain of ChlD, we show that this site is essential for magnesium chelation and allosterically regulates Mg(2+) and MgATP(2-) binding.