PloS one
Targeted re-sequencing identified rs3106189 at the 5' UTR of TAPBP and rs1052918 at the 3' UTR of TCF3 to be associated with the overall survival of colorectal cancer patients.
PMID 23940558
Abstract
Recent studies have demonstrated the power of deep re-sequencing of the whole genome or exome in understanding cancer genomes. However, targeted capture of selected genomic whole gene-body regions, rather than the whole exome, have several advantages: 1) the genes can be selected based on biology or a hypothesis; 2) mutations in promoter and intronic regions, which have important regulatory roles, can be investigated; and 3) less expensive than whole genome or whole exome sequencing. Therefore, we designed custom high-density oligonucleotide microarrays (NimbleGen Inc.) to capture approximately 1.7 Mb target regions comprising the genomic regions of 28 genes related to colorectal cancer including genes belonging to the WNT signaling pathway, as well as important transcription factors or colon-specific genes that are over expressed in colorectal cancer (CRC). The 1.7 Mb targeted regions were sequenced with a coverage ranged from 32× to 45× for the 28 genes. We identified a total of 2342 sequence variations in the CRC and corresponding adjacent normal tissues. Among them, 738 were novel sequence variations based on comparisons with the SNP database (dbSNP135). We validated 56 of 66 SNPs in a separate cohort of 30 CRC tissues using Sequenom MassARRAY iPLEX Platform, suggesting a validation rate of at least 85% (56/66). We found 15 missense mutations among the exonic variations, 21 synonymous SNPs that were predicted to change the exonic splicing motifs, 31 UTR SNPs that were predicted to occur at the transcription factor binding sites, 20 intronic SNPs located near the splicing sites, 43 SNPs in conserved transcription factor binding sites and 32 in CpG islands. Finally, we determined that rs3106189, localized to the 5' UTR of antigen presenting tapasin binding protein (TAPBP), and rs1052918, localized to the 3' UTR of transcription factor 3 (TCF3), were associated with overall survival of CRC patients.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AV45437
Anti-TAPBP antibody produced in rabbit, affinity isolated antibody