EMAIL THIS PAGE TO A FRIEND
BMC Developmental Biology
Changes in sub-cellular localisation of trophoblast and inner cell mass specific transcription factors during bovine preimplantation development.
PMID 23941255