EMAIL THIS PAGE TO A FRIEND
Dermatologic Surgery : Official Publication For American Society For Dermatologic Surgery [Et Al.]
A review of the metabolism of 1,4-butanediol diglycidyl ether-crosslinked hyaluronic acid dermal fillers.
PMID 23941624