EMAIL THIS PAGE TO A FRIEND
Emergency medicine journal : EMJ
Best BETs from the Manchester Royal Infirmary. BET 1: intranasal lorazepam is an acceptable alternative to intravenous lorazepam in the control of acute seizures in children.
PMID 23943640
Abstract
A short-cut review was carried out to determine whether intranasal lorazepam was as effective as intravenous lorazepam in the control of seizures in children. Eighteen papers were found using the reported search, of which one was directly relevant and another compared intranasal lorazepam with intramuscular paraldehyde. The author, date and country of publication, patient group studied, study type, relevant outcomes, results and study weaknesses are shown in table 1. It is concluded that intranasal lorazepam appears to be a safe and effective treatment for this condition.