EMAIL THIS PAGE TO A FRIEND
Water Environment Research : A Research Publication of the Water Environment Federation
Performance evaluation and modeling of synthetic-fiber barrier in the treatment of turbid water.
PMID 23944142