EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Correlation between functionality preference of Ru carbenes and exo/endo product selectivity for clarifying the mechanism of ring-closing enyne metathesis.
PMID 23944769
Abstract
Functionality preferences of metathesis Ru carbenes to various alkenes and alkynes with electronic and steric diversity were determined by using time-dependent fluorescence quenching. The functionality preferences depend not only on the properties of multiple bonds but also on the ligands on Ru. There was a good correlation between functionality preference and the metathesis reaction outcome. The correlation between functionality preference and exo/endo product ratio offers a solution to resolve the mechanistic issue related with alkene- vs alkyne-initiated pathway in ring-closing enyne metathesis. The correlation indicates the preference is likely to dictate the reaction pathway and eventually the outcome of the reaction. The Ru catalyst favoring alkyne over alkene provides more endo product, indicating that the reaction mainly initiates at the alkyne. By changing the substitution pattern, the preference can be reversed to give an exclusive exo product.