EMAIL THIS PAGE TO A FRIEND
Drug development and industrial pharmacy
Characterization of micellar systems produced by new amphiphilic conjugates of poly(ethylene glycol).
PMID 23944836
Abstract
This study proposes polymeric micelles produced using new amphiphilic conjugates between amino- or carboxy-mPEG2000 and three different α-lipoamino acids (PEG-LAA). The characterization of these colloidal systems showed CMC values, in the order of 10(-5 )M, that are interesting in the view of an in vivo administration. The PEG-LAA micelles also showed a good stability at 37 °C and upon dilution in aqueous media. Using a colored probe as a model lipophilic compound, the loading efficiency and in vitro release profile were also outlined.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
94425
Propionic acid, analytical standard
C3H6O2