EMAIL THIS PAGE TO A FRIEND
Chimia
Synthesis and reactivity of Zn-biphenyl metal-organic frameworks, introducing a diphenylphosphino functional group.
PMID 23945099
Abstract
Structural features, synthesis, and reactivity of Zn-biphenyl metal-organic frameworks with MOF-5 topology are presented to show the chemical flexibility of such materials and to demonstrate the challenges that can be encountered and solved to avoid interpenetration. We introduce the synthesis of a Zn-biphenyl MOF with diphenylphosphino functionalization and illustrate its structural and chemical properties.