EMAIL THIS PAGE TO A FRIEND
Clinical immunology (Orlando, Fla.)
Impaired Th1 immunity in ovarian cancer patients is mediated by TNFR2+ Tregs within the tumor microenvironment.
PMID 23948613
Abstract
Ovarian cancer is a prevalent gynecological malignancy with potent immune-suppression capabilities; regulatory T cells (Tregs) are significant contributors to this immune-suppression. As ovarian cancer patients present with high levels of TNF and Tregs expressing TNFR2 are associated with maximal suppressive capacity, we investigated TNFR2+ Tregs within these patients. Indeed, TNFR2+ Tregs from tumor-associated ascites were the most potent suppressor T cell fraction. They were abundantly present within the ascites and more suppressive than peripheral blood TNFR2+ Tregs in patients. The increased suppressive capacity can be explained by a distinct cell surface expression profile, which includes high levels of CD39, CD73, TGF-β and GARP. Additionally, CD73 expression level on TNFR2+ Tregs was inversely correlated with IFN-γ production by effector T cells. This Treg fraction can be selectively recruited into the ascites from the peripheral blood of patients. Targeting TNFR2+ Tregs may offer new approaches to enhance the poor survival rates of ovarian cancer.