EMAIL THIS PAGE TO A FRIEND
Organic letters
The synthesis of indoline and benzofuran scaffolds using a Suzuki-Miyaura coupling/oxidative cyclization strategy.
PMID 23952242
Abstract
The generation of indolines and benzofurans from the combination of Suzuki-Miyaura coupling reactions with oxidative cyclizations is described.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I5605
Indoline, ReagentPlus®, 99%
C8H9N