EMAIL THIS PAGE TO A FRIEND
Talanta
Identification of selenium-containing proteins in HEK 293 kidney cells using multiple chromatographies, LC-ICPMS and nano-LC-ESIMS.
PMID 23953436
Abstract
Our previous studies using HeLa and HEK 293 cells demonstrated that selenomethionine, SeMet, exerts more of an antagonistic effect on arsenic than other selenium species. These studies attributed the antagonistic effect of SeMet to decreased levels of reactive oxygen species, ROS, changes in protein phosphorylation and possible incorporation of SeMet into proteins. The present study employs a metallomics approach to identify the selenium-containing proteins in HEK 293 cells raised with SeMet. The proteins were screened and separated using two dimensional high performance liquid chromatography (HPLC)-inductively coupled plasma mass spectrometry (ICPMS), size exclusion chromatography (SEC) and reversed-phase chromatography (RPC). The Se-containing proteins were identified by peptide mapping using nano-HPLC-Chip-electrospray ionization mass spectrometry (ESIMS).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S3875
Seleno-DL-methionine, ≥99% (TLC)
C5H11NO2Se
S3132
Seleno-L-methionine, ≥98% (TLC), powder
C5H11NO2Se