EMAIL THIS PAGE TO A FRIEND
Current atherosclerosis reports
Cadmium exposure and clinical cardiovascular disease: a systematic review.
PMID 23955722
Abstract
Mounting evidence supports that cadmium, a toxic metal found in tobacco, air and food, is a cardiovascular risk factor. Our objective was to conduct a systematic review of epidemiologic studies evaluating the association between cadmium exposure and cardiovascular disease. Twelve studies were identified. Overall, the pooled relative risks (95% confidence interval) for cardiovascular disease, coronary heart disease, stroke, and peripheral arterial disease were: 1.36 (95% CI: 1.11, 1.66), 1.30 (95% CI: 1.12, 1.52), 1.18 (95% CI: 0.86, 1.59), and 1.49 (95% CI: 1.15, 1.92), respectively. The pooled relative risks for cardiovascular disease in men, women and never smokers were 1.29 (1.12, 1.48), 1.20 (0.92, 1.56) and 1.27 (0.97, 1.67), respectively. Together with experimental evidence, our review supports the association between cadmium exposure and cardiovascular disease, especially for coronary heart disease. The number of studies with stroke, heart failure (HF) and peripheral arterial disease (PAD) endpoints was small. More studies, especially studies evaluating incident endpoints, are needed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GF61336997
Cadmium, wire reel, 5m, diameter 0.5mm, hard, 99.99+%
Cd
GF88672786
Cadmium, wire reel, 5m, diameter 0.125mm, as drawn, 99.9%
Cd
GF97910950
Cadmium, wire reel, 0.1m, diameter 0.125mm, as drawn, 99.9%
Cd
GF42263849
Cadmium, wire reel, 0.1m, diameter 1.0mm, hard, 99.999%
Cd
GF12224078
Cadmium, wire reel, 0.2m, diameter 0.25mm, hard, 99.99+%
Cd
GF23019960
Cadmium, wire reel, 0.2m, diameter 0.5mm, hard, 99.95+%
Cd
GF37213893
Cadmium, wire reel, 0.2m, diameter 0.5mm, hard, 99.99+%
Cd
GF37063144
Cadmium, wire reel, 0.2m, diameter 0.70mm, as drawn, 99.99+%
Cd
GF80208536
Cadmium, wire reel, 0.2m, diameter 1.2mm, as drawn, 99.99+%
Cd
GF39849935
Cadmium, wire reel, 0.2m, diameter 2.0mm, hard, 99.99+%
Cd
GF38869105
Cadmium, wire reel, 0.2m, diameter 0.125mm, as drawn, 99.9%
Cd
GF82981064
Cadmium, wire reel, 0.2m, diameter 1.0mm, hard, 99.999%
Cd
GF28009593
Cadmium, wire reel, 0.5m, diameter 0.25mm, hard, 99.99+%
Cd
GF77495833
Cadmium, wire reel, 0.5m, diameter 0.5mm, hard, 99.95+%
Cd
GF51736653
Cadmium, wire reel, 0.5m, diameter 0.5mm, hard, 99.99+%
Cd
GF12560868
Cadmium, wire reel, 0.5m, diameter 0.70mm, as drawn, 99.99+%
Cd
GF55395298
Cadmium, wire reel, 0.5m, diameter 1.2mm, as drawn, 99.99+%
Cd
GF81241523
Cadmium, wire reel, 0.5m, diameter 2.0mm, hard, 99.99+%
Cd
GF58637762
Cadmium, wire reel, 0.5m, diameter 0.125mm, as drawn, 99.9%
Cd
GF72632998
Cadmium, wire reel, 0.5m, diameter 1.0mm, hard, 99.999%
Cd
GF30757383
Cadmium, wire reel, 10m, diameter 0.25mm, hard, 99.99+%
Cd
GF01198762
Cadmium, wire reel, 1m, diameter 0.25mm, hard, 99.99+%
Cd
GF86979986
Cadmium, wire reel, 1m, diameter 0.5mm, hard, 99.95+%
Cd
GF67082328
Cadmium, wire reel, 1m, diameter 0.5mm, hard, 99.99+%
Cd
GF63053276
Cadmium, wire reel, 1m, diameter 0.70mm, as drawn, 99.99+%
Cd
GF01474635
Cadmium, wire reel, 1m, diameter 1.2mm, as drawn, 99.99+%
Cd
GF29720875
Cadmium, wire reel, 1m, diameter 0.125mm, as drawn, 99.9%
Cd
GF79124441
Cadmium, wire reel, 1m, diameter 1.0mm, hard, 99.999%
Cd
GF72498347
Cadmium, wire reel, 2m, diameter 0.5mm, hard, 99.95+%
Cd
GF85290699
Cadmium, wire reel, 2m, diameter 0.5mm, hard, 99.99+%
Cd