EMAIL THIS PAGE TO A FRIEND
Journal of the American Academy of Dermatology
Efficacy and safety of calcipotriol/betamethasone dipropionate ointment for the treatment of simple onycholysis: an open-label study.
PMID 23957979