EMAIL THIS PAGE TO A FRIEND
Human reproduction update
The periconceptional period, reproduction and long-term health of offspring: the importance of one-carbon metabolism.
PMID 23959022
Abstract
BACKGROUND Most reproductive failures originate during the periconceptional period and are influenced by the age and the lifestyle of parents-to-be. We advance the hypothesis that these failures can arise as a partial consequence of derangements to one-carbon (1-C) metabolism (i.e. metabolic pathways that utilize substrates/cofactors such as methionine, vitamin B12, folate). 1-C metabolic pathways drive the synthesis of proteins, biogenic amines and lipids required for early growth, together with the synthesis and methylation of DNA and histones essential for the regulation of gene expression. We review how deficiencies in periconceptional 1-C metabolism affect fertility and development together with underlying mechanisms derived from animal studies. METHODS A literature search was performed using PubMed and bibliographies of all relevant original research articles and reviews. RESULTS We define 'periconception' as a 5-6-month period in women embracing oocyte growth, fertilization, conceptus formation and development to Week 10 of gestation (coinciding with the closure of the secondary palate in the embryo). During this period significant epigenetic modifications to chromatin occur that correspond with normal development. Subtle variations in 1-C metabolism genes and deficiencies in 1-C substrates/cofactors together with poor lifestyle, such as smoking and alcohol consumption, disturb 1-C metabolism and contribute to subfertility and early miscarriage and compromise offspring health. Procedures used in assisted reproduction can also disturb these metabolic pathways and contribute to poor pregnancy outcomes. CONCLUSIONS Evidence presented indicates that parental nutrition and other lifestyle factors during the periconceptional period can affect reproductive performance via 1-C metabolic pathways. This knowledge provides opportunities for treatment and prevention of reproductive failures and future non-communicable diseases.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C9157
Activated charcoal, suitable for cell culture, suitable for plant cell culture
C
C2764
Activated charcoal, untreated, granular, ≤5 mm
C
C2889
Activated charcoal, untreated, granular, 8-20 mesh
C
161551
Activated charcoal, powder, -100 particle size (mesh), decolorizing
C
292591
Activated charcoal, 4-14 mesh, granular, Norit® PK 3-5
C
242233
Activated charcoal, DARCO®, 4-12 mesh particle size, granular
C
242241
Activated charcoal, DARCO®, 12-20 mesh, granular
C
242268
Activated charcoal, DARCO®, 20-40 mesh particle size, granular
C
242276
Activated charcoal, DARCO®, −100 mesh particle size, powder
C
329428
Activated charcoal, NORIT® ROW 0.8 SUPRA, pellets
C
C3014
Activated charcoal, untreated, granular, 20-60 mesh
C
C4386
Activated charcoal, acid-washed with hydrochloric acid
C
C5510
Activated charcoal, acid-washed with phosphoric and sulfuric acids
C
05112
Activated charcoal, for GC
C
05105
Activated charcoal, powder
C
31616
Activated charcoal, puriss. p.a., powder
C
675326
Activated charcoal, DARCO® KB-G
C
80485
Activated charcoal, for the determination of AOX, 50-150 μm particle size
C
C7606
Activated charcoal, meets USP testing specifications
C
97876
Activated Charcoal Norit®, Norit CA1, from wood, chemically activated, powder
C
37771
Activated Charcoal Norit®, Norit GAC 1240W, from coal, for potable water processing, steam activated, granular
C
22874
Activated Charcoal Norit®, Norit PK 1-3, from peat, steam activated, granular
C
29204
Activated Charcoal Norit®, Norit RB3, for gas purification, steam activated, rod
C
51127
Activated Charcoal Norit®, Norit RB4C, steam activated, rod, for the purification of gas
C
53663
Activated Charcoal Norit®, Norit SX ultra, from peat, corresponds U.S. Food chemicals codex (3rd Ed.), steam activated and acid washed, highly purified, powder
C
93067
Activated Charcoal Norit®, Norit® SX2, powder, from peat, multi-purpose activated charcoal, steam activated and acid washed
C
484164 Carbon, glassy, spherical powder, 2-12 μm, 99.95% trace metals basis
C
633100 Carbon, nanopowder, <100 nm particle size (TEM)
C
GF29093058
Carbon, chopped fiber, 100g, nominal diameter 0.007mm, fiber length 6mm, grade 34-700
C
GF02887342
Carbon, fabric, 1000x1000mm, weight.muü 375g, thickness 0.4mm, ends x picks/10cm 47x47, 2/2 twill weave
C