EMAIL THIS PAGE TO A FRIEND
Circulation
Response to letters regarding article, “Randomized comparison of sevoflurane versus propofol to reduce perioperative myocardial ischemia in patients undergoing noncardiac surgery".
PMID 23961559